Learn More
Cayman Chemical AM251, 183232-66-8, 10 mg
SDP
Supplier:  Cayman Chemical 7167010
Encompass_Preferred
A CB1 antagonist and CB2 inverse agonist; Kis = 7.5 and 2,290 nM for CB1 and CB2 from rat forebrain and mouse spleen, respectively; agonist at GPR55 (EC50 = 39 nM); potentiates GABAA currents (EC50 = 0.4 µM); prevents TGF-β1-induced EMT in HK-2 renal tubule epithelial cells independent of CB receptors or GPR55; halts the cell cycle and induces apoptosis in A375 human melanoma cells; reduces antidepressant-like behavior and decreases fasting-induced hyperphagia in mice
Catalog No. NC9004721
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.