Learn More
Apexbio Technology LLC AM966,5mg CAS# 1228690-19-4
SDP
Supplier:  Apexbio Technology LLC A31705
Encompass_Preferred
AM966 is a selective, potent and orally bioavailable antagonist of lysophosphatidic acid type 1 receptor (LPA1) with IC50 values of 17 and 19 nM for human or mouse LPA1, respectively [1]. Relative to LPA2, 3, 4, 5 receptors, AM966 shows 10-fold and 100-fold more selective in mouse and human cell lines for LPA1, respectively [1].In vitro, AM966 shows to inhibit LPA-induced chemotaxis in IMR-90 human lung fibroblasts, A2058 human melanoma cells and CHO cells expressing LPA1 receptors [1].In vivo, AM966 has been demonstrated to reduce total BALF cells, LDH activity, BALF collagen and total TGFβ concentrations in bleomycin-induced mice. Moreover, AM966 has been reported to block lung fibrosis, reduce lung inflammation and maintain body weight in mice induced by bleomycin [1]. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-224-0471
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.