Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Amber Glass Reagent Bottle with Screw Cap
SCI_ED

Catalog No. S40891 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S40891 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S40891 Supplier Eisco™ Supplier No. CH0159C
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Amber Glass Reagent Bottle with Screw Cap made from soda glass. Amber color protects photosensitive materials from UV light.

  • Economy amber reagent bottle
  • Made with amber soda glass
  • Plastic screw cap
  • 4 in. height
  • 2 in. bottle diameter
  • 0.75 in. diameter bottle opening

Warranty and Services

1 year limited manufacturer warranty.

Specifications

Capacity (Metric) 125 mL
Color Amber
Description Reagent Bottles
Diameter (English) 2 in.
Diameter (English) Inner Neck 0.7 in.
Diameter (Metric) Inner Neck 17.78 mm
For Use With (Application) Holding liquids
Includes Screw cap
Material Soda Lime Glass
Length (Metric) 5 cm
Quantity 1 Ea.
Sterility Non-sterile
Weight (Metric) 9 g
Width (Metric) 5 cm
Closure Color White
Closure Included Yes
Graduated No
Mouth Narrow
Height (Metric) 11.4 cm
Height (English) 4.6 in.
Chemical Label Blank
Closure Size 2 cm
Diameter (Metric) 5 cm
Closure Type Screw
Autoclavable Autoclavable
Capacity (English) 3.38 oz.
Shape Cylindrical
Disposable Reusable
Product Type Reagent Bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.