Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Amber Molded Glass Type I Borosilicate Serum Vials

Catalog No. 01672068
Change view
Click to view available options
Volume (Metric):
10 mL
20 mL
50 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
01-672-068 10 mL
01-672-072 50 mL
01-672-073 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-672-068 Supplier Qorpak™ Supplier No. 327205
Add to Cart
Add to Cart

The utilization of type I molded glass enhances mechanical resilience, thereby minimizing the potential for particle contamination during crucial fill/finish and lyophilization stages. Vials are depyrogenated and sterilized. Crimp cap not included.

  • 100% inspected in clean room environment
  • Vials are washed with water for injection and dried in an ISO8 environment
  • Vials are depyrogenated in a tunnel according to European and US pharmaccopeias
  • Vials are placed in trays for packaging in ISO5 area with ISO7 surrounding
  • Sealed with a Tyvek™ lid in ISO5 environment, packed in steribags and placed into abox allowing for sterilization in ISO8 surrounding
  • Sterilization is made through ethylene oxide and a process validated according to the ISO11135 normal

Specifications

Color Amber
Diameter (Metric) Outer 25.4 mm
Material Borosilicate Glass
Pre-slit No
Sterility Sterile
Disposable Single-use
Volume (Metric) 10 mL
For Use With (Application) Lyophilization
Graduated No
No. per Case 640/Cs.
No. per Pack 80/Pk.
Packaging Type Tray Packed Neck Up
Size (Metric) Opening 20 mm
Dnase Free No
Closure Type Crimp Cap
Quantity 640 Vials
Marking Spot No
Fit for Purpose Sterile
Vial Type Ready to Use/Sterile
Autoclavable Autoclavable
Height (Metric) 53.5 mm
Product Type Serum Vial
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.