Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco Labs Amber Volumetric Flask with Polyethylene Stopper and QR Code
SCI_ED

Catalog No. S01906 Shop All Eisco Products
Change view
Click to view available options
Capacity (Metric):
5 mL
10 mL
20 mL
25 mL
50 mL
100 mL
200 mL
250 mL
500 mL
1000 mL
2000 mL
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S01906 5 mL
S01907 10 mL
S000593 20 mL
S01903 25 mL
S000592 50 mL
S01905 100 mL
S01898 200 mL
S01899 250 mL
S01900 500 mL
S01901 1000 mL
S01902 2000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. S01906 Supplier Eisco Labs Supplier No. CH3041
Add to Cart
Add to Cart

Class A Amber Volumetric Flask is made from durable borosilicate glass, has a secure polyethylene stopper, and features a QR code for certification — ideal for precise lab work and safe handling of light-sensitive solutions.

Amber Volumetric Flask is made from borosilicate 3.3 glass and includes an interchangeable polyethylene stopper with a joint for secure closure. It features a QR Code for checking certifications.

  • Amber-colored glass reduces light transmission, suitable for use with photosensitive contents during storage and handling in light-exposed environments.
  • Class A tolerance, conforms to ASTM E1042 standards; calibrated to contain (TC) with permanently marked QR code for tracking.
  • Suitable for solution preparation, titration, and volume-sensitive dilution tasks in chemical, analytical, or pharmaceutical labs.
  • Resistant to thermal stress, chemical exposure, and autoclaving at 121°C; low expansion 3.3 borosilicate glass supports long-term use.

Specifications

Color Amber
Capacity (Metric) 5 mL
Material Borosilicate Glass (3.3)
Product Type Volumetric Flask
Tolerance ±0.025 mL
Autoclavable Not Autoclavable
Disposable Reusable
Graduated No
Mouth Style Narrow
Sterility Non-sterile
Bottom Shape Flat
Closure Size 10/19
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.