Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ AmberChrom™ (formerly Dowex™) 50WX4 Hydrogen Form, 200-400 mesh

Catalog No. 111015222 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5222 2.5 kg
11-101-5228 100 g
11-101-5215 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-101-5222 Supplier MilliporeSigma™ Supelco™ Supplier No. 2174842.5KG
Add to Cart
Add to Cart

This strongly acidic cation exchange resin has not been specially processed nor cleaned. It is suggested that it be sujbected to a suitable preliminary elution and wash.

Dowex™ 50WX4 hydrogen form is a nonmacroporous gelform bead resin. Dowex™ 50WX4 hydrogen form is a useful column packing material, used in anion exchange chromatography.
TRUSTED_SUSTAINABILITY

Specifications

Particle Size 200/400 Mesh
Packaging Poly Bottle
Quantity 2.5 kg
Product Line Amberchrom
Capacity (Metric) 1.1 meq/mL by wetted bed volume
MDL Number MFCD00132725
CAS 69011-20-7

Amberchrom is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.