Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amberlite IRA-68, ion exchange resin, Thermo Scientific™

Catalog No. p-7023566
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1725322 100 g
AAA1725336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1725322 Supplier Thermo Scientific Chemicals Supplier No. A1725322

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It finds its application as a weakly basic anion exchange resin with an unusually high capacity for large organics. It is also applied in deacidification, deionization of process liquors, isolation of acidic natural products, purification of novabiacin and cephalosporins, separation of neutral and acidic amino acids below ph 10, removal of heparinic acid at low ph, concentration of citric acid, and demineralization of cheese whey.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9056-59-1
MDL Number MFCD00145795

Specifications

Odor Amine-like
Merck Index 14,382
Quantity 100 g
Chemical Name or Material Amberlite IRA-68, ion exchange resin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.