Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CellsDirect Resuspension & Lysis Buffers includes 10 ml Resuspension Buffer & 1 ml Lysis Buffer

Catalog No. 11739010
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-739-010 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-739-010 Supplier Invitrogen™ Supplier No. 11739010
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Includes

10mL Resuspension Buffer, 1mL Lysis Buffer

These two buffers are to be used only as a complement to the SuperScript™ III Platinum™ CellsDirect Kits when large number of samples are processed.

These two buffers are to be used only as a complement to the SuperScript™ III Platinum™ CellsDirect Kits when large number of samples are processed. See manual of SuperScript III Platinum CellsDirect Kits for protocol recommendations.

  • Fewer resources used, less hazardous, less waste

Order Info

Shipping Condition: Dry ice

TRUSTED_SUSTAINABILITY

Specifications

Concentration 2X
Content And Storage Store in freezer (-5 to -30°C).
Product Line CellsDirect
Product Type Cell Lysis Buffer
Quantity 1 kit
Shipping Condition Dry Ice
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.