Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Conical Tubes (50 mL) (racked)

Catalog No. AM12501
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 tubes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM12501 200 tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AM12501 Supplier Invitrogen™ Supplier No. AM12501
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Invitrogen RNase-free Conical Tubes are provided assembled with green screw caps and guaranteed RNase- and DNase-free.

  • Provided assembled with green screw caps
  • The tubes have a printed writing space and volume marks on the sidewalls for convenience
  • These polypropylene tubes are sterile, nuclease-free, nonpyrogenic, and can be autoclaved to 121°C
  • 200 x 50 mL tubes

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 50 mL
Material Polypropylene
Quantity 200 tubes
Sterility Sterile
Content And Storage Store at room temperature.
Product Line Ambion
Shipping Condition Room Temperature
RNase Free Yes
Dnase Free Yes
Autoclavable Autoclavable
Shape Conical
Bottom Shape Conical
Closure Type Screw Cap
Closure Included Yes
Writing Area Yes
Product Type Conical Tube
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.