Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNase-Free Elution Tubes (2.0 mL)

Catalog No. AM12480
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tubes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM12480 100 Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AM12480 Supplier Invitrogen™ Supplier No. AM12480
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

These elution tubes are guaranteed RNase- and DNase-free.

Invitrogen microcentrifuge tubes are guaranteed RNase- and DNase-free.

Features include:

  • Marked with graduations for convenience
  • Flat lids for easy labeling
  • Positive seal design that prevents caps from opening in boiling water baths
  • Autoclave-safe

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Capacity (Metric) 2 mL
Material Polypropylene
Quantity 100 Tubes
Content And Storage Store at room temperature.
Product Line Invitrogen
Shipping Condition Room Temperature
Dnase Free RNase-Free, DNase-Free
Autoclavable Autoclavable
Closure Type Snap Cap
Graduated Yes
Product Type Elution Tube
Surface Type Graduated
Show More Show Less
Can the collection tubes within the MEGAclear Transcription Clean-Up Kit (Cat. No. AM1908) be ordered separately?

Yes, the Elution Tubes (2.0 mL) (Cat. No. AM12480) are the same that are used in the MEGAclear Transcription Clean-Up Kit (Cat. No. AM1908).

WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.