Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ambion™ RNase III

Catalog No. AM2290
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 units
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM2290 250 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AM2290 Supplier Invitrogen™ Supplier No. AM2290
Get 19% Off Invitrogen Molecular Biology Reagents
Use promo code INV19
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Double-stranded RNA (dsRNA) specific endoribonuclease

Escherichia coli Ribonuclease III (RNase III; EC 3.1.24) is specific endoribonuclease. In E. coli, RNase III cleaves ribosomal RNA (rRNA) precursors during maturation of rRNA. Enzyme cleaves dsRNA into 12–15 bp dsRNA fragments with 2 to 3 nucleotide 3' overhangs, and 5' phosphate and 3' hydroxyl termini. Termini and overhangs of RNase III cleavage products are thus same as those produced by Dicer in eukaryotic RNAi pathway. Transfection of RNase III cleavage products can be used to induce RNAi in mammalian cells (patent pending).

  • Unit Definition: One unit is defined as amount of enzyme catalyzing the cleavage of 1μg of 500 bp dsRNA substrate to approximately 12–30 bp fragments in 60 min. at 37°C

DNA & RNA Purification & Analysis, In Vitro Transcribed siRNA & Dicer siRNA, Nuclease Protection Assays, Nucleic Acid Gel Electrophoresis & Blotting, RNAi, RNAi, Epigenetics & Noncoding RNA Research

Order Info

Shipping Conditions: Dry ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage RNase III and 10X Reaction Buffer should be stored at –20°C.
Shipping Condition Dry Ice
Enzyme RNase
Compatible Buffer 10X Reaction Buffer
Quantity 250 units
Product Type RNase III
Product Line Ambion

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.