Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ambion™ SP6 RNA Polymerase, cloned, 20 U/μL

Catalog No. AM2071
Encompass_Preferred
Change view
Click to view available options
Quantity:
1,000 units, 20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM2071 1,000 units, 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AM2071 Supplier Invitrogen™ Supplier No. AM2071
Get 19% Off Invitrogen Molecular Biology Reagents
Use promo code INV19
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Includes

One tube containing 1000 units at a concentration of 20U/μL

A cloned high-purity RNA polymerase with a high specificity for its promoter

 

  • This enzyme will transcribe large amounts of RNA from DNA sequences downstream of the promoter and show no cross talk between promoters
  • One tube containing 1000U at a concentration of 20U/μL is provided. Its high purity makes it ideal for synthesizing high specific activity RNA hybridization probes, biologically active mRNA, and antisense RNA
  • The RNA Polymerase is rigorously tested for contaminating nonspecific endonuclease, exonuclease, protease, and RNase activity
  • Functionality of enzyme is tested in In Vitro Transcription reaction under optimal and limiting UTP concentration.

Unit Definition: One unit is the amount of SP6 RNA Polymerase required to catalyze the incorporation of 1 nmol of nucleoside triphosphate in 60 minutes at 37°C

 

DNA & RNA Purification & Analysis, Gene Expression Analysis & Genotyping, in vitro Transcription, Nucleic Acid Labeling & Oligo Synthesis, RNA Labeling

Order Info

Shipping Condition: Dry ice

TRUSTED_SUSTAINABILITY

Specifications

Polymerase SP6 RNA Polymerase
Quantity 1,000 units, 20 μL
Concentration 20 U/μL
Source DNA

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.