Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ULTRAhyb™-Oligo

Catalog No. AM8663
Encompass_Preferred
Change view
Click to view available options
Quantity:
125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM8663 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AM8663 Supplier Invitrogen™ Supplier No. AM8663
Get 19% Off Invitrogen Molecular Biology Reagents
Use promo code INV19
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifically formulated for use with oligonucleotide probes in northern analysis

  • Contains a proprietary blend of hybridization accelerators and blocking agents to enhance the levels of hybridization
  • Supplied in one bottle containing 125mL
  • Use less probe per blot
  • Decrease hybridization time to 2 hours
  • Using standard hybridization buffers, only 1 to 5% of target molecules on a blot hybridizes to probe, making blots a relatively insensitive method for nucleic acid analysis (Vernier et al. (1996) Anal Biochem 235: 11-19)
  • With ULTRAhyb™ Ultrasensitive Hybridization Buffer, the hybridization reaction approaches completion, so that as few as 10,000 molecules can be detected
  • It works well with oligonucleotides having Tms from 78° to 90°C and 47 to 62% GC contents; However, the optimal hybridization temperature for oligonucleotide probes with different GC content and Tm values should be determined empirically

DNA and RNA Purification and Analysis, Northern Blotting, Nucleic Acid Gel Electrophoresis and Blotting

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at 4°C.
Buffer Hybridization Buffer
Quantity 125 mL
Shipping Condition Room Temperature
Product Type Oligo Buffer
What membranes are compatible with your ULTRAhyb-Oligo solution?

ULTRAhyb-Oligo solution contains 25% formamide and is only compatible with positively charged or neutral nylon membranes.

With the ULTRAhyb-Oligo Hybridization Buffer, why am I getting cross-hybridization?

The extreme sensitivity of ULTRAhyb-Oligo Hybridization Buffer may allow detection of RNAs that are not the expected full-length target. Although the probe binding could be legitimate (hybridization to alternatively spliced, partially degraded, or closely related mRNAs), some might be hybridization to RNAs with only partial homology to the target. The easiest way to reduce signal from cross hybridization (either cause) is to simply reduce the exposure time.
Here are other possible causes for cross-hybridization and solutions offered:
- Hybridization stringency may be inadequate. We recommend increasing the hybridization temperature by 2-5 degrees C and increasing the wash temperature by 5-10 degrees C.
- The probe contains non-target sequence. If the oligonucleotide has sequence homology with other mRNAs, vectors, etc., we recommend redesigning the probe to avoid sequence homology with targets other than the intended target.

With the ULTRAhyb-Oligo Hybridization Buffer, why am I getting low signal on my blot?

Here are possible causes and solutions:
- Not enough probe was used or low specific activity probe was used. The amount and specific activity of probe needed to obtain radioactive signal above background depends largely on the amount of target on the blot and the specific activity of the labeled probe. Maximize signal by using a molar excess of probe labeled to the highest possible specific activity.

- Hybridization and/or washes are too stringent. If the oligonucleotide has a very low GC content or a low Tm, then lowering the hybridization temperature by 2-5 degrees C may increase the signal by reducing the hybridization stringency. Note that this may increase background and cross-hybridization. Alternatively washing can be done at room temperature instead of at 42 degrees C.

With the ULTRAhyb-Oligo Hybridization Buffer, I am getting high background on my blot. Why is this?

Here are possible causes and solutions:
- There could be precipitates in the ULTRAhyb-Oligo Hybridization Buffer. Inadequate solubilization of the hybridization buffer is one of the primary causes of high background. While warming the buffer at 68 degrees C, we recommend thoroughly mixing it with a gentle swirling motion. Ensure that there is no precipitate in the buffer before adding it to the blot. ULTRAhyb-Oligo Hybridization Buffer may start to precipitate at temperatures below 25-30 degrees C.

- Prehybridization may have been inadequate. We recommend increasing the blot prehybridization time from 30 min to 1 hr.

- Unincorporated radionucleotides may have been present. We recommend that free label be removed with a spin column (i.e., Ambion NucAway Spin Columns, Cat. No. AM10070) before adding the oligonucleotide probe to the hybridization solution.

- Washing may have been inadequate. We recommend the following:
-Ensure that your wash buffer contains SDS. Wash buffers lacking SDS are not recommended for use with ULTRAhyb-Oligo Hybridization Buffer.
-Do the post-hybridization washes in 5X SSC or SSPE, 0.5% SDS for 2 x 30 min. Increasing the salt concentration will help remove probe that is non-specifically bound to the membrane through electrostatic interactions.
-Double the post-hybridization wash time in 2X SSC or SSPE, 0.5% SDS from 2 x 30 min to 4 x 30 min.
-If none of the previous suggestions reduce non-specific background sufficiently, try increasing the post-hybridization wash temperature by 5-10 degrees C. This should be the last resort as it may remove hybridized probe in addition to reducing background.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.