Learn More
TARGETMOL CHEMICALS INC Ambrisentan 50MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T165550MG
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Ambrisentan (BSF 208075) is an endothelin receptor antagonist used in the therapy of pulmonary arterial hypertension (PAH). Ambrisentan has been associated with a low rate of serum enzyme elevations during therapy but has yet to be implicated in cases of clinically apparent acute liver injury. Purity 99.86%
Catalog No. 50-287-7378
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.