Learn More
Enzo Life Sciences AMC (7-Amino-4-methylcoumarin) Standard (1 ml) CAS: 26093-31-2
SDP
Supplier:  Enzo Life Sciences BMLKI1070001
Encompass_Preferred
Fluorogenic chromophore. Ex(max): 380nm, Em(max): 460nm. Reference compound. Formula: C10H9NO2. MW: 175.2. Formulation: 30µM in 50mM HEPES, pH 7.4, 100mM sodium chloride, 0.1% CHAPS, 10mM dithiothreitol, 1mM EDTA, 10% glycerol.
Catalog No. 50-210-0525
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.