Learn More
AdipoGen AMCA-H N-succinimidyl ester
SDP
Supplier:  AdipoGen CDXA0010M025
Encompass_Preferred
Chemical. CAS 113721-87-2. Formula C16H14N2O6. MW 330.29. Synthetic. AMCA-H NHS ester reacts with primary amine groups on antibodies, proteins, peptides and other biomolecules at pH 7.0-9.0. Suitable for labeling lysyl residues in proteins Ex/Em 353/455nm.
Catalog No. 50-256-1872
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.