Learn More
AdipoGen AMCA-H
SDP
Supplier:  AdipoGen CDXA0009G002
Encompass_Preferred
Chemical. CAS 106562-32-7. Formula C12H11NO4. MW 233.22. Synthetic. AMCA is one of the brightest amine-reactive blue fluorescent dyes useful for immunofluorescence and fluorescent labeling Ex/Em 353/455nm. It is quite photostable, and its fluorescence is pH-independent from pH 4 to 10. The properties include a relatively large Stokes shift and resistance to photobleaching. Reactive AMCA derivatives are used as contrasting probes for double and triple labeling in immunofluorescence microscopy, arrays and in situ hybridization. NHS-AMCA and Sulfo-NHS-AMCA are reactive towards primary amine groups on antibodies, proteins, peptides and other biomolecules. AMCA-Hydrazide is used to label glycosylation sites or for conjugation to carboxyl groups using the crosslinker EDC.
Catalog No. 50-256-1875
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.