Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ AMD 3100 octahydrochloride
GREENER_CHOICE

Catalog No. p-4783738
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-991-0 10 mg
32-995-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 32-991-0 Supplier Tocris Bioscience™ Supplier No. 3299/10
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Highly selective CXCR4 antagonist

Highly selective CXCR4 chemokine receptor antagonist (IC50 values are 0.02 - 0.13 and > 25 μM for CXCR4 and all other chemokine receptors respectively). Switches inflammatory responses from Th2 to Th1 type and reduces airway hyperresponsiveness in a mouse model of asthma. Potently inhibits HIV-1 and HIV-2 replication in vitro (EC50 = 4 - 35 nM) and mobilizes hematopoietic stem cells in vivo. Attenuates cocaine place preference and locomotor stimulation in rats. Attenuates microglial activation neurological function after ischemic stroke in mice.

Chemical Identifiers

CAS 155148-31-5
Molecular Formula C28H62Cl8N8

Specifications

Quantity 10 mg
Formula Weight 794.48
Purity >98%
Molecular Weight (g/mol) 794.46
Chemical Name or Material AMD 3100 octahydrochloride
Recommended Storage Desiccate at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.