Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AMDEX Goat Anti-Rabbit IgG - Horseradish Peroxidase, 0.5ml Secondary Antibody, Cytiva

Catalog No. 45001175 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-001-175 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-001-175 Supplier Cytiva Supplier No. RPN4301
Add to Cart
Add to Cart

Host Species: Goat
Isotype: IgG
Species Reactivity: Rb

Enzyme Immunoassay

TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG
Classification Monoclonal
Conjugate HRP
Description Goat Anti-Rabbit IgG - Horseradish Peroxidase
Dilution For ELISA, it is recommended that the conjugate should be diluted in the range 1:1000 to 1:4000
Host Species Goat
Purification Method Affinity Purified
Quantity 0.5 mL
Primary or Secondary Secondary
Isotype IgG
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.