Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Amdex Streptavidin-HRP Enzyme Conjugate

Catalog No. 45001177 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-001-177 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-001-177 Supplier Cytiva Supplier No. RPN4401
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For ELISA signal amplification. The conjugates are based on novel chemistry which facilitates conjugation of multiple enzyme molecules (alkaline phosphatase or horseradish peroxidase) to a single streptavidin or antibody molecule via a dextran backbone.

The conjugates are based on novel chemistry which facilitates conjugation of multiple enzyme molecules (alkaline phosphatase or horseradish peroxidase) to a single streptavidin or antibody molecule via a dextran backbone.

  • 10- to 100-fold increase in signal intensity
  • Compatible with traditional ELISA systems
  • Option to reduce incubation times
  • Well controlled nonspecific binding properties

TRUSTED_SUSTAINABILITY

Specifications

Conjugate HRP
Quantity 0.5 mL
Sample Volume 500 μL
Content And Storage 2°C to 8°C
Form Liquid
For Use With (Application) For use in ELISA systems
Buffer Stabilized in 50% TRIS Buffer and 50% HRP Conjugate Stabilizer
Product Type Strepavidin HRP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.