Learn More
American Educational Products Aneroid Sphygmomanometer
SCI_ED
Catalog No. S97324 Shop All American Educational Products LLC Products
Change view
Click to view available options
Aneroid Sphygmomanometer
Catalog No.
S97324 Aneroid Sphygmomanometer
1 options
This item is intended for educational purposes only.
Catalog No. S97324 Supplier American Educational Products Supplier No. 71371
This item is intended for educational purposes only.
Used to check blood pressure.
  • Comprised by an inflatable cuff to restrict blood flow, and a mercury or mechanical manometer to measure the pressure
  • Used in conjunction with a means to determine at what pressure blood flow is just starting, and at what pressure it is unimpeded
Specifications
For Use With (Application) Used to check blood pressure
Includes Inflatable cuff and a mercury or mechanical manometer
Product Type Aneroid Sphygmomanometer