Learn More
Invitrogen™ AMER2 Polyclonal Antibody

Description
Immunogen sequence: VDDTYLQEFW DMLSQTEEQG PEPQEGAAKV AAALETKVVP ETPKDTRCVE AAKDASSVKR RRLNRIPIEP HPKEEPKHPE KEQQE Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 74%.
Specifications
Specifications
| Antigen | AMER2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | AMER2 |
| Gene Accession No. | Q8N7J2 |
| Gene Alias | 2600011E07Rik; AMER2; APC membrane recruitment 2; APC membrane recruitment protein 2; Fam123a; family with sequence similarity 123, member A; family with sequence similarity 123A; protein FAM123A; RGD1559605 |
| Gene Symbols | AMER2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.