Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Amerex™ Halotron I Clean-Agent Stored-Pressure Fire Extinguishers
Most widely used Halon 1211 replacement agent
$950.46
Specifications
Product Type | Fire Extinguisher |
---|---|
Material | Steel |
Discharge Time | 9 sec. |
Reagent Type | Halotron™ I |
Temperature Range (English) | -40° to +120°F |
Catalog Number | Mfr. No. | Includes | Model | Range | UL Rating | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | Model | Range | UL Rating | Price | Quantity & Availability | ||
19-035-394
![]() |
Amerex™
15271 |
Anodized Aluminum valve, nozzle, vehicle bracket | B386T | 9 to 15 ft. | 5B:C |
Each of 1 for $950.46
|
|
||
Description
- Halotron I is a Clean Agent hydrochlorofluorocarbon (HCFC) discharged as a rapidly evaporating liquid which leaves no residue
- Effectively extinguishes Class A and B fires by cooling and smothering; will not conduct electricity back to the operator
- Pressurized with argon gas, an EPA and FAA approved agent suitable for use on Class A, B and C fires
Warranty and Services
Five years.
Specifications
Fire Extinguisher | |
9 sec. | |
-40° to +120°F |
Steel | |
Halotron™ I | |
-40°C to +48.8°C |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title