Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

American Educational Products Dropping Bottles
SCI_ED

Catalog No. S31688 Shop All American Educational Products LLC Products
Change view
Click to view available options
Capacity (Metric):
15 mL
30 mL
250 mL
Color:
Amber
Clear
Shape:
French Square
Round
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Color Shape
S31688 250 mL Amber Round
S31671 15 mL Clear French Square
S31672 30 mL Clear Round
S31682 30 mL Amber Round
S31678 250 mL Clear Round
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. S31688 Supplier American Educational Products Supplier No. 7S31688

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Store reagent solutions in these sturdy dropping bottles.

  • Come with Bakelite™ screw tops, glass pipets and rubber bulbs
  • Available in French Square or Boston Round shape, clear or amber color and a wide range of sizes

Order Info

Replacement pipets available for each bottle (See Cat. No. S31671P).

Specifications

Capacity 8.45 oz., 250 mL
Capacity (Metric) 250 mL
Color Amber
Includes Dropper assembly
Material Glass
Closure Type Screw Cap
Capacity (English) 8.45 oz.
Shape Round
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.