Learn More
American Educational Products Electrical Bench Meters
SCI_ED
Catalog No. S77842 Shop All American Educational Products LLC Products
Change view
Click to view available options
Product Type:
Ammeter, Bench DC
Voltmeter, Bench
Amperage:
0 to 5 A
Scale:
Single
Voltage:
0 to 5 V
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Amperage Scale Voltage
S77842 Ammeter, Bench DC 0 to 5 A Single -
S77811 Voltmeter, Bench - Single 0 to 5 V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S77842 Supplier American Educational Products Supplier No. 7130910
Designed in a wide array of options to fit your needs.
  • Low-cost,  high-quality  meters  for  numerous  uses
  • Closed back
Specifications
For Use With (Application) Ideal for student use
Quantity Each
Product Type Ammeter, Bench DC
Style Closed Back
Includes Insulated thumbscrew
Scale Single
Amperage 0 to 5 A