Learn More
American Educational Products Lung Demonstration Apparatus
SCI_ED
Catalog No. S41390 Shop All American Educational Products LLC Products
Change view
Click to view available options
Demonstration apparatus
Catalog No.
S41390 Demonstration apparatus
1 options
This item is not returnable. View return policy
Catalog No. S41390 Supplier American Educational Products Supplier No. 712033
This item is not returnable. View return policy
Relaxation and contraction illustrated.
This simple model demonstrates the mechanisms of breathing for students.
  • Shows how the lungs are emptied and filled with air by the relaxation and contraction of the diaphragm muscle
  • One or two balloons attached to a Y-tube simulate the lungs
  • The chest cavity is a half-gallon bell jar with open top and stout rubber sheet that acts as the diaphragm
  • As the diaphragm is pulled downward, the atmospheric pressure is lowered inside the chamber, causing the lungs to expand and take in air
  • When the diaphragm is released, the lungs deflate
Specifications
For Use With (Application) Simple model demonstrates the mechanisms of breathing for students
Material Plastic
Product Type Lung Replacement Rubber Dam