Learn More
                                    Andwin Scientific Citrus Clearing Solvent 3.8L
Supplier: Andwin Scientific 72060044
            Citrus Clearing Solvent 3.8L. D-limonene based citrus solvent has a slight citrus scent and a higher flash point eliminates the need for storage in a flammable safety cabinet. Highly effective xylene substitute for manual or automated systems.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.