Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Amersham 2'-Deoxyuridine 5'-Triphosphate Solution

Catalog No. 01183281 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
100 μmol
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-183-281 100 μmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 01-183-281 Supplier Cytiva Supplier No. 28406542
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Amersham high purity dNTPs for DNA amplification, dideoxy sequencing, labelling, mutagenesis, cDNA synthesis, and protein expression profiling.

  • Cytiva dNTPs offer the highest purity deoxynucleotides available for DNA amplification, dideoxy sequencing, labelling, mutagenesis, complementary DNA (cDNA) synthesis, and protein expression profiling.
    • Free from DNase, RNase, and nicking enzyme activity.
    • Buffer free and ready-to-use solutions in multiple formats.
    • Custom formulation, testing, packaging, and labeling are available.
  • Functionally tested for long Polymerase Chain Reaction (PCR) and sequencing, with greater than 99% triphosphate purity (by HPLC) for optimum performance and consistency.
TRUSTED_SUSTAINABILITY

Specifications

Format Liquid
Product Type dNTPs
Concentration 100 mM
Purity > 99% Triphosphate
Quantity 100 μmol
Color Colorless
For Use With (Application) DNA Amplification, Dideoxy Sequencing, Labelling, Mutagenesis, cDNA Synthesis, and Protein Expression Profiling
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.