Learn More
Cytiva Amersham™ AlkPhos Direct™ ECF Detection Module
GSA_VA
Catalog No. 45000938 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
Each
Catalog No. Quantity
45-000-938 Each
1 options
Catalog No. 45-000-938 Supplier Cytiva Supplier No. RPN3685
Add to Cart
Add to Cart
Fast and convenient systems for all high-sensitivity applications using DNA, RNA or oligonucleotide probes
AlkPhos Direct Labeling and Detection Systems are based on the rapid, direct labeling of DNA or RNA probes with thermostable alkaline phosphatase. CDP-Star™ or ECF™ detection can be used. DNA or RNA probes are labeled during a 30-minute reaction and resulting probes can be used without purification. Hybridization stringency can be controlled by temperature as well as salt concentration.
  • Hybridization stringency can be controlled by elevating temperatures up to 75°C
  • Detection of 60 fg target DNA (single-copy gene) in a genomic Southern blot
  • ECF detection for quantitation experiments
  • Rapid protocol saves two to three hours over conventional indirect methods
Order Info
Each system include the following reagents, sufficient for 25 or 50 labeling reactions and detection of 2500cm2: labeling reagent, crosslinker, control DNA, reaction buffer, hybridization buffer, blocking agent, ECF detection reagents.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Sufficient For 2500cm2 membrane
Includes Sufficient for 25 or 50 labeling reactions and detection of 2500cm2 membrane: labeling reagent, crosslinker, control DNA, reaction buffer, hybridization buffer, blocking agent, ECF detection reagents.
Quantity Each