Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Amersham™ Cy™3, Cy™ 5 Maleimide mono-Reactive Dye 5-packs

Catalog No. 45001273 Shop All Cytiva Products
Change view
Click to view available options
Label or Dye:
Cy3
Cy5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Label or Dye
45-001-273 Cy3
45-001-274 Cy5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45-001-273 Supplier Cytiva Supplier No. PA23031
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

More selective method of antibody labeling that reduces the possibility of the label interfering with subsequent antibody-antigen reactions. Amersham™ Cy™3, Cy™ 5 Maleimide mono-Reactive Dye packs are packaged in premeasured amounts and foil-sealed to ensure consistent labelings.

Cy3 and Cy5 mono-functional maleimides are particularly suitable for the selective labeling of molecules containing free sulfhydryl groups, such as cysteine residues in proteins and peptides, and oligonucleotides. They are also useful in antibody labeling since the dye molecules are introduced away from the antibody binding site, increasing the chance that functionality of the labeled molecule will be unimpaired. The protocol is simple and robust, offering an alternative chemistry when either greater selectivity of labeling is required or amine labeling with Cy3 and Cy5 NHS esters is not desirable or appropriate.

  • Provides an alternative for when CyDye™ NHS esters are unsuitable.
  • Dyes are packaged in premeasured amounts and foil-sealed to ensure consistent labelings

Order Info

Each kit contains five vials of dye, each sufficient for labeling 1mg of protein.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) For the labelling of biological compounds with Cy3 monofunctional dye
Quantity 5 vials each sufficient to label 1 mg protein
Product Type Cy3 Maleimide Mono-Reactive Dye 5-Pack
Kit Contents Five vials of dye
Sufficient For Labeling 1mg Protein
Molecular Weight (g/mol) 791
Solubility Soluble in cold water and hot water
Content And Storage 2°C to 8°C, protected from light. Reconstituted material may be stored for up to 2 weeks at -20°C in aliquots to avoid repeat freeze thaw cycles.
Label or Dye Cy3
Color Red
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.