Learn More
Description
Fluorescence detection is a direct Western blotting method in which a secondary antibody is conjugated to a fluorophore, avoiding the need for ancillary detection reagents. Different fluorophores are available for signal detection at either visible light or near-infrared (NIR) wavelengths.
Amersham CyDye 700 and 800 secondary antibodies are labeled with NIR fluorophores that emit light at wavelengths of 700 nm or 800 nm. By using Amersham CyDye 700 and 800 secondary antibodies together with a pair of appropriate rabbit or mouse primary antibodies, you can perform multiplexed experiments.
Key benefits:
- Easy handling and a strong signal-to-noise ratio yield reproducible results
- Multiplexing analysis of more than one target protein on a single blot increases process efficiency
- Total protein normalization (TPN) generates reliable and accurate quantitation
- Stable signals allow membranes to be stored and rescanned for future studies
Specifications
Specifications
| Applications | Western Blot |
| Conjugate | CyDye 800 |
| Quantity | 0.1 mg |
| Primary or Secondary | Secondary |
| Content And Storage | 2°C to 8°C, protected from light |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.