Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Amersham™ CyDye™ Antibody Labeling Kits

Catalog No. 45000726 Shop All Cytiva Products
Change view
Click to view available options
Label or Dye:
Cy3 Reactive Dye
Cy5 Reactive Dye
Product Type:
Cy3 Ab Labeling Kit
Cy5 Ab Labeling Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Label or Dye Product Type
45-000-726 Cy5 Reactive Dye Cy5 Ab Labeling Kit
01-183-290 Cy3 Reactive Dye Cy3 Ab Labeling Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45-000-726 Supplier Cytiva Supplier No. PA35000

missing translation for 'validMainframeMsg'

Convenient way to tag antibodies with CyDye fluorescent dyes

CyDye fluorescent dyes are supplied as highly water soluble, reactive NHS esters for easy conjugation to biomolecules containing aliphatic amino groups.

  • Conjugations are carried out under mild conditions and are complete within 1 hour
  • Dyes are packaged in premeasured amounts and foil sealed to ensure consistent labelings

Order Info

Antibody kits contain all reagents required for 2 x 1mg reactions. Monoclonal antibody kits contain all reagents required for 2 x 0.1mg reactions.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) For the conjugation of proteins with Cy5 Reactive Dye
Quantity Each
Product Type Cy5 Ab Labeling Kit
pH Range 7 to 7.3
Molecular Weight (g/mol) 975.15
Solubility Cy5 dye: Soluble in cold water and hot water Coupling buffer: Soluble in cold water and hot water Elution buffer: Soluble in cold water and hot water
Content And Storage 2°C to 8°C, protected from light
Label or Dye Cy5 Reactive Dye
Color Blue
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.