Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Amersham CyDye DIGE Fluor Dye, Cy3, Cy5
GSA_VA

Catalog No. 45002404 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
600 nmol
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-002-404 600 nmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-002-404 Supplier Cytiva Supplier No. 25800984

missing translation for 'validMainframeMsg'

Designed for use in 2-D Fluorescence Difference Gel Electrophoresis using the Ettan DIGE system. Cytiva Amersham CyDye DIGE Fluor minimal dyes and saturation dyes are size- and charge-matched fluorescent dyes specifically engineered for detecting protein abundance differences.

CyDye DIGE fluors are exceptional dyes for multicolor analysis, offering bright and intense colors with narrow excitation and emission bands. The fluors are spectrally distinct, making them highly suitable for multicolor detection. CyDye DIGE fluors utilize these benefits, but are also size- and charge-matched specifically for 2-D DIGE using the Ettan DIGE system.
  • Allows detection of up to three prelabeled protein samples and standards on the same 2-D electrophoresis gel
  • Size- and charge-matched dyes enable co-migration of labeled samples within the gel
  • Bright and highly sensitive dyes allow the use of the minimal labeling technique
  • Minimal loss of signal during labeling, separation, and scanning
  • No change in signal over wide pH range used during first-dimension (IEF) separation
  • Discrete signal from each fluor with minimal cross-talk contributes to high accuracy
Available as minimal and saturation labeling dyes
  • The minimal dyes are intended for general 2-D application use where sufficient amounts of sample are available
  • The saturation dyes from the Scarce Sample Labeling Kit are designed to be used for applications where only small amounts of sample are available, for example in Laser Capture Microdissection

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 600 nmol
Kit Contents 100 nmol CyDye DIGE fluor Cy3 saturation dye for analytical labeling
100 nmol CyDye DIGE fluor Cy5 saturation dye for analytical labeling
400 nmol CyDye DIGE fluor Cy3 saturation dye for preparative labeling
Content And Storage -15°C to -30°C , Avoid light, store in the dark.
Label or Dye Cy3, Cy5
Color Red, Blue
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.