Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
For labeling DNA and RNA probes especially for multicolor analysis in microarrays, FISH, chromosome identification, whole chromosome painting, karyotyping, and gene mapping. CyDye labeled nucleotides are available in a range of bright, intense colors with narrow emission bands—ideal for multiplexing within a single sample.
- Low nonspecific binding
- High photostability
- High water solubility
- pH insensitive
Specifications
Specifications
| Description | CyDyes are a group of bright fluorescent dyes which can be used in genomic and protein research. Cy dye labelled nucleotides exhibit the following properties: |
| pH | 7 |
| Molecular Weight (g/mol) | 791.99 (Monofunction), 975.15 (Bisfunction) |
| Product Type | Cy5-dCTP |
| Reagents | Fluorescent Labeling Reagents |
| Wavelength | 670nm |
| Content And Storage | -15°C to -30°C |
| Quantity | 25 nmol |
| Color | Far-Red |
| For Use With (Application) | Genomic and Protein research, labeling DNA and RNA probes, especially for multicolor analysis in microarrays, FISH, chromosome identification, whole chromosome painting, karyotyping and gene mapping |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?