Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Amersham™ CyDye™ Reactive Dye 5-Packs

Catalog No. p-3676017 Shop All Cytiva Products
Change view
Click to view available options
Label or Dye:
Cy3 mono-Functional Dye
Cy5 mono-Functional Dye
Cy5.5 mono-Functional Dye
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Label or Dye
45-000-719 Cy5 mono-Functional Dye
45-000-715 Cy3 mono-Functional Dye
01-183-289 Cy5.5 mono-Functional Dye
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 45-000-719 Supplier Cytiva Supplier No. PA25001
Only null left
Add to Cart
Add to Cart

Use for the most accurate labeling. Cytiva Amersham™ CyDye™ Reactive Dye 5-Packs contain derivatized CyDye that carries only one reactive group on each dye molecule for accurate labeling of amine groups.

 

    • Convenient way to label antibodies, proteins, drugs, and oligonucleotides with CyDye Fluorescent Dyes.
    • For use in ELISA, immunoprecipitation, and blotting applications.
    • Dyes are packaged in premixed amounts and foil-sealed to ensure consistent labelings.

 

Order Info

Each kit contains five vials of dye. Each vial is sufficient for labeling 1mg protein or 30nmol amino groups.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) For the labelling of biological compounds with Cy5 monofunctional dye
Quantity Each
Product Type Cy5 Mono-Reactive Dye Pack
Kit Contents Five foil packs each containing dried dye to label 1mg of Protein
Sufficient For Labeling 1mg Protein or 30nM Amino groups
Molecular Weight (g/mol) 791.99
Content And Storage 2°C to 8°C, protected from light
Label or Dye Cy5 mono-Functional Dye
Color Blue
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.