Learn More
Description
Hydrazide esters provide a convenient method for the labeling of free carbonyl groups of glycoproteins and carbohydrates. For maximum convenience, stable active hydrazide esters are supplied and stored as solid material.
- Range of bulk packs available for greatest economy
- Example protocols included with each dye pack
Specifications
Specifications
| For Use With (Application) | For labelling via carbonyl groups. For use in steady state fluorescence, FP and FRET applications |
| Quantity | 1 mg |
| Product Type | Cy5.5 Hydrazide |
| pH Range | 3 to 10 |
| Molecular Weight (g/mol) | 1128.42 |
| Solubility | Soluble in cold water and hot water |
| Content And Storage | 2°C to 8°C, protected from light. Reconstituted material may be stored for up to 2 weeks at -20°C in aliquots to avoid repeat freeze thaw cycles. |
| Label or Dye | Cy5.5 |
| Color | Dark Blue |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.