Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Amersham™ ECF™ Substrate for Western Blotting
GSA_VA

Catalog No. 45000947 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
Sufficient reagents for detecting 2500cm2 membrane
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-000-947 Sufficient reagents for detecting 2500cm2 membrane
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 45-000-947 Supplier Cytiva Supplier No. RPN5785
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

ECF Substrate, 60mL; Dilution buffer

Used for detection of protein blots with fluorescence scanning equipment. Cytiva Life Sciences™ Amersham™ ECF™ Substrate for Western Blotting uses a fluorescent substrate for alkaline phosphatase-based detection of protein blots.

Alkaline phosphatase dephosphorylates ECF Substrate to produce a fluorescent product, which localizes at the site of catalysis. Product fluoresces at 540 to 560nm when excited at the appropriate wavelength.

  • For use with fluorescence scanning equipment
  • Sensitivity comparable to chemiluminescence
  • Wide dynamic range of fluorescence scanning systems compared with film allows visualization and quantitation of weak and dark bands in the same scan
  • Eliminate need for darkroom and film

Western blotting, dot, and slot blotting

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Fluorescence scanning equipment
Includes ECF Substrate, 60mL, Dilution Buffer
Quantity Sufficient reagents for detecting 2500cm2 membrane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.