Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Amersham™ ECL™ Direct Nucleic Acid Labeling/Detection System

Catalog No. p-3676034 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
For 2000 cm2 Membrane
For 4000 cm2 membrane
Labels 10 μg Nucleic Acid
Labels 5 μg Nucleic Acid
Product Type:
Detection Reagents
Hybridization Buffer
Labeling Module
Labeling and Detection System
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
45-000-925 Labels 5 μg Nucleic Acid Labeling Module
45-000-874 For 4000 cm2 membrane Detection Reagents
45-000-923 Labels 10 μg Nucleic Acid Labeling and Detection System
45-000-924 For 2000 cm2 Membrane Detection Reagents
45-000-922 Labels 5 μg Nucleic Acid Labeling and Detection System
45-000-926 For 4000 cm2 membrane Hybridization Buffer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 45-000-925 Supplier Cytiva Supplier No. RPN3005
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

45-000-922 and 45-000-923: Labeling reagent; Crosslinker; Control DNA; Blocking agent; ECL detection reagents and ECL Gold hybridization buffer

Use for fast and easy detection of medium-to-high-target amounts. Cytiva Life Sciences™ Amersham™ ECL™ Direct Nucleic Acid Labeling/Detection System are based on the direct labeling of DNA or RNA probes with horseradish peroxidase (HRP) in a simple 20 min chemical reaction

 

    • Direct probe labeling in a 10 min reaction, 1 h from hybridization to detection with ECL Direct, Hybond N+, and Hyperfilm ECL.
    • Eliminates handling, waste, and regulatory issues associated with the use of radioactivity.
    • No need to strip blots before reprobing.
    • For fast and easy detection of medium- to high-target amounts in applications such as colony/plaque screens, dot blots, and PCR product analyses.
    • Consistent results combining strong signals with very low backgrounds.

 

 

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
For Use With (Application) Colony or plaque screens, dot blots and PCR product analyses
Includes DNA labelling reagent 0.5mL; Glutaraldehyde solution 0.5mL; Water 1.0mL; l Hind III control DNA (10μg/mL) 0.1 ml; Gold hybridization buffer 500mL; Blocking agent 25g
Product Type Labeling Module
Quantity Labels 5 μg Nucleic Acid
Test Time 20 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.