Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Amersham™ ECL™ Plex Fluorescent Rainbow Markers
GSA_VA

Catalog No. 45001595 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
120 μL
500 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-001-595 500 μL
45-001-594 120 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45-001-595 Supplier Cytiva Supplier No. RPN851E
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Optimized for use with the ECL Plex Western Blotting Detection System. Amersham™ ECL™ Plex Fluorescent Rainbow Markers are ideal for quantitative western blot.

  • Monitor progress of protein electrophoresis and assess transfer efficiency and molecular weight of blotted proteins without staining
  • Enhanced to enable faster and simpler identification of proteins on SDS-polyacrylamide gels
  • Ready-to-load
  • Provide sharper, more intense bands on gels and blots and have improved band spacing for more accurate molecular weight determination
  • Provides fluorescent images using Cy3 and Cy5 channels

Order Info

Each 120μL pack size is sufficient for use with at least 40 minigels (10 x 8cm) or 20 large gels (20 x 20cm). Each 500μL pack size is sufficient for use with at least 160 minigels (10 x 8cm) or 80 large gels (20 x 20cm).

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Description These fluorescent rainbow markers are for use with the ECL Plex Western blotting detection system.
Color Multi
Formulation Supplied in 35% Glycerol and sample buffer containing Mercaptoethanesulphonic Acid (MESNA) as reducing agent
For Use With (Application) For quantitative western blots
Quantity 500 μL
Storage Requirements -20°C
Sufficient For 50 Minigel Loadings
Product Type Protein Molecular Weight Marker
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.