Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Amersham™ ECL Plex™ Western Blotting Combination Packs

Catalog No. 45001215 Shop All Cytiva Products
Change view
Click to view available options
Formulation:
Cy3, Cy5, Amersham Hybond LFP 0.2 PVDF
Cy3, Cy5, Amersham Protran Premium 0.45 NC
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation
45-001-215 Cy3, Cy5, Amersham Hybond LFP 0.2 PVDF
45-001-214 Cy3, Cy5, Amersham Protran Premium 0.45 NC
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45-001-215 Supplier Cytiva Supplier No. RPN999
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Amersham ECL Plex goat-α-mouse IgG-Cy3, 150μg; Amersham ECL Plex goat-α-rabbit IgG-Cy5, 150μg; Amersham ECL Plex Fluorescent Rainbow Markers, 120μL; 45-001-214: Amersham Protran Premium, 10 x 10cm, 10 sheets or 45-001-215: Hybond LFP 0.2μM PVDF, 20 x 20cm, 3 sheets

Complete system optimized for fluorescent Western blotting

  • Improved formulation of Cy3 conjugates for even lower background

TRUSTED_SUSTAINABILITY

Specifications

Formulation Cy3, Cy5, Amersham Hybond LFP 0.2 PVDF
For Use With (Application) ECL Plex western blotting system
Kit Contents ECL Plex Goat-α-mouse IgG-Cy3 150μg
ECL Plex Goat-α-rabbit IgG-Cy5 150μg
ECL Plex Fluorescent Rainbow Markers 120μL
Hybond LFP, 10 Sheets (20 x 20cm)
Quantity 1 Kit: ECL Plex Goat-α-mouse IgG-Cy3: 150 μg, ECL Plex Goat-α-rabbit IgG-Cy5: 150 μg, ECL Plex Fluorescent Rainbow Markers: 120 μL, Hybond LFP: 10 Sheets (20 x 20cm)
Storage Requirements Varies by component
Product Type Western Blot Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.