Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Amersham™ Rediprime II DNA Labeling System

Catalog No. 45000812 Shop All Cytiva Products
Change view
Click to view available options
30 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
45-000-812 30 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-000-812 Supplier Cytiva Supplier No. RPN1633

missing translation for 'validMainframeMsg'

Long shelf-life and simple, fast protocol. Amersham™ Rediprime II DNA Labeling System consists of premixed, individually dispensed, random-prime reactions, freeze-dried for ambient stability.

p>Simply add substrate and [α-32P]dCTP. As little as 25ng substrate DNA can be used.

  • Ambient temperature-stable reactions with a color check for the labeling of DNA to specific activities of >109dpm/μg
  • Long shelf life and simple, fast protocol; labeling reaction complete in 10 min.
  • No need to remove unincorporated nucleotides before hybridization
  • DNA can be labeled in solution or in the presence of low melting point agarose

Suitable for use with dCTP only.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 25°C
For Use With (Application) Labelling of DNA from a variety of sources to a high specific activity for use in Southern and Northern blot hybridizations
Includes Labeling reaction Buffered solution of dATP, dGTP, dTTP, Exonuclease-free Klenow Enzyme and random primers in a dried, stabilized form, control DNA 300ng of lambda Hind III DNA in a dried, stabilized form 1 tube
No. of Reactions 30
Product Type Rediprime II DNA Labeling System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.