Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Amersham™ Streptavidin-Biotinylated HRP Complex

Catalog No. 45000749 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-000-749 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-000-749 Supplier Cytiva Supplier No. RPN10512ML
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Streptavidin conjugated to reporter enzyme molecules for use in blotting and in situ hybridization applications

  • High specificity reagents
  • Conjugation achieved using the periodate coupling method described by Nakane and co-workers
  • Sold on the basis of titer in an ELISA and performance in a western blot
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Suitable for use in ECL™, western blotting ,ELISA, immunocytochemistry
Includes 2mL of Phosphate Buffered Saline (0.1M Sodium Phosphate, 0.1M Sodium Chloride, pH 7.5) containing 1%(w/v) Bovine Serum Albumin and an antimicrobial agent
Quantity 2 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.