Learn More
Cytiva Amersham™ Streptavidin-Fluor Conjugates
Catalog No. 45000734 Shop All Cytiva Products
Change view
Click to view available options
Product Type:
Cy3-Streptavidin
Cy5-Streptavidin
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
45-000-734 Cy5-Streptavidin
45-000-731 Cy3-Streptavidin
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45-000-734 Supplier Cytiva Supplier No. PA45001
  • PRODUCT PROMOTION
    $436.50
    $394.86 / Each of 1
    Save $41.64
    10% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Add to Cart
Use for in situ hybridization and flow cytometry. Cytiva Life Sciences™ Amersham™ Streptavidin-Fluor Conjugates consist of streptavidin conjugated to a range of fluorescent dyes.
  • High specificity reagents
Applications:
in situ hybridization, flow cytometry
TRUSTED_SUSTAINABILITY
Specifications
Formulation This product is supplied as a lyophilized solid in Phosphate-buffered Saline (0.02M Potassium Phosphate, 0.15M NaCl) pH 7.2, containing 1%(w/v) Bovine Serum Albumin and 0.1% (w/v) Sodium Azide, reconstitute with 1mL of deionized Water, dispense into suitable aliquots and freeze, avoid repeated freeze-thaw cycles
For Use With (Application) Suitable for use in the Fluorescent analysis of Biotinylated samples
Quantity 1 mg
Product Type Cy5-Streptavidin
Kit Contents Lyophilized solid in phosphate-buffered saline (0.02M potassium phosphate, 0.15M NaCl) pH 7.2, containing 1%(w/v) bovine serum albumin and 0.1% (w/v) sodium azide, reconstitute with 1mL of deionized water, dispense into suitable aliquots and freeze