Learn More
United States Biological Corporation Ames Medium w/L-Glutamine, Powder, 50 L
Catalog No. NC1068474 Shop All United States Biological Corporation Products
Encompass_Preferred
Ames' medium was formulated to support retinal tissue in relatively short-term culture. Rabbit retina has been incubated in Ames' medium for over 2 days with its metabolism and electrical responses to light stimuli well maintained. This mixture is a medium of choice for maintaining central nervous system tissue in vitro.
Catalog No. NC1068474 Supplier United States Biological Corporation Supplier No. A13722550L
Ames' medium was formulated to support retinal tissue in relatively short-term culture. Rabbit retina has been incubated in Ames' medium for over 2 days with its metabolism and electrical responses to light stimuli well maintained. This mixture is a medium of choice for maintaining central nervous system tissue in vitro.