Learn More
TARGETMOL CHEMICALS INC AMG-0347 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T29968
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AMG-0347 (UNII-CD7L9290QR) is a small molecule inhibitor targeting TRPA1(transient receptor potential ankyrin 1) ion channels a receptor found in sensory neurons and involved in the detection of pain and environmental stimuli. AMG-0347 acts by blocking the function of TRPA1 and has analgesic effects. purity: 99%
Catalog No. 50-258-0441
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.