Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ AMG 517
GREENER_CHOICE

Catalog No. 599510
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
59-951-0 10 mg
59-955-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 59-951-0 Supplier Tocris Bioscience™ Supplier No. 5995/10
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent TRPV1 antagonist

Potent TRPV1 antagonist (IC50 = 0.9 nM). Inhibits capsaican-induced flinching and reverses analgesia in an inflammatory pain model in rats. Orally bioavailable.

Chemical Identifiers

CAS 659730-32-2
Molecular Formula C20H13F3N4O2S

Specifications

Quantity 10 mg
Solubility Soluble to 100mM in DMSO
Formula Weight 430.41
Purity >98%
Molecular Weight (g/mol) 430.405
Chemical Name or Material N-[4-[[6-[4-(Trifluoromethyl)phenyl]-4-pyrimidinyl]oxy]-2-benzothiazolyl]acetamide
Recommended Storage Store at 4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.