Learn More
Apexbio Technology LLC AZD1480 935666-88-9 10mg
SDP
Supplier:  Apexbio Technology LLC A413710
Encompass_Preferred
AZD1480 (CAS 935666-88-9) is a small molecule inhibitor targeting Janus kinase (JAK) showing selectivity for JAK2 over JAK3 and modest selectivity over JAK1 at physiological ATP concentrations By suppressing phosphorylation-mediated activation of JAK2 AZD1480 subsequently inhibits downstream STAT3 and FGFR3 signaling pathways involved in tumor cell proliferation angiogenesis and metastasis Preclinical studies demonstrate its capability to reduce cellular growth and induce apoptosis in multiple myeloma cell lines (e g RPMI 8226 OPM-2 NCI-H929 Kms 18) primary myeloma samples and mouse xenograft tumor models AZD1480 serves as a probe in research investigating JAK-STAT signal transduction and cancer biology
Catalog No. 50-000-62564
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.