Learn More
Ansh Labs AMH (Rat and Mouse) ELISA 96-wells, 50 L Plasma/Serum, Range 2 calibrators ranging 0.23-15 ng/mL, LOD TBD, Time 3.5 hours, Shelf Life 24 months
SDP
Supplier:  Ansh Labs AL113
Encompass_Preferred
Anti-Mullerian Hormone AMH (Rat and Mouse) ELISA has a total 3.5 hours incubation, calibrators and range of 2, 0.313-20 ng/mL, sample of only 5 µL predilution Serum. Reacts with Camel, Mouse, Rat. Crossreactivity with related molecules is not significant.
Catalog No. 50-194-3726
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.