Learn More
Apexbio Technology LLC MEK162 (ARRY-162, ARRY-438162) 606143-89-9 50mg
Supplier: Apexbio Technology LLC A194750
MEK162 (ARRY-162 ARRY-438162 CAS 606143-89-9) is a selective inhibitor of MEK1 and MEK2 kinases exhibiting an IC50 of approximately 12 nM By targeting the MAPK/ERK signaling pathway MEK162 inhibits ERK1/2 phosphorylation reduces clonal survival and induces apoptosis in melanoma cell models including those harboring N-RAS GNAQ or GNA11 mutations Studies also indicate that combining MEK162 with PKC inhibitors leads to sustained suppression of MAPK signaling and enhanced pro-apoptotic effects in uveal melanoma cells Early-phase clinical research has reported transient typically mild retinopathy associated with MEK162 treatment This compound is widely utilized in studies of targeted melanoma therapies
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.