Learn More
Enzo Life Sciences AMI-1 (5mg). CAS: 20324-87-2
SDP
Supplier:  Enzo Life Sciences ALX270440M005
Encompass_Preferred
Potent, cell permeable and specific inhibitor of protein arginine N-methyltransferases (PRMTs) (IC50=8.8µM for human PRMT1 and 3.0µM for yeast Hmt1p). Does not compete for the AdoMet (S-adenosyl-L-methionine; SAM) binding site. Exhibits only minimal effects on lysine N-methyltransferases. Suppresses the transcriptional coactivator activity of PRMT1 and PRMT4. Inhibitor of HIV-1 RT polymerase (IC50=5.0µM). Purity: ≥98% (HPLC). Appearance: Brown solid. Solubility: Soluble in water, 100% ethanol or DMSO. Long Term Storage: -20°C.
Catalog No. 50-200-8561
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.